4'-Bromo-3-chloro-4-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68534-1G 68534-25G 68534-500MG 68534-5G
76935-788EA 380.87 USD
76935-788 76935-790 76935-792 76935-794
4'-Bromo-3-chloro-4-fluoro-1,1'-biphenyl ≥97%
4'-Bromo-3-chloro-4-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇BrClF
MW: 285.539 g/mol
Storage Temperature: Ambient
CAS Number: 1500884-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR