5-(Benzyloxy)-2-chloro-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

68182-1G 68182-25G 68182-500MG 68182-5G
76935-772EA 215.37 USD
76935-772 76935-774 76935-776 76935-778
5-(Benzyloxy)-2-chloro-3-methylpyridine ≥97%
5-(Benzyloxy)-2-chloro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNO
Storage Temperature: Ambient
CAS Number: 74650-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR