4-Chloro-2-fluoro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67874-1G 67874-25G 67874-500MG 67874-5G
76935-758EA 317.21 USD
76935-758 76935-760 76935-762 76935-764
4-Chloro-2-fluoro-5-methoxybenzoic acid ≥97%
4-Chloro-2-fluoro-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
Storage Temperature: Ambient
CAS Number: 146447-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR