4-Chloro-3-fluoro-2-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

66520-1G 66520-500MG 66520-5G
76935-746EA 797.8 USD
76935-746 76935-748 76935-750
4-Chloro-3-fluoro-2-methoxybenzamide ≥97%
4-Chloro-3-fluoro-2-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₇ClFNO₂
MW: 203.598 g/mol
Storage Temperature: Ambient
CAS Number: 2387172-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR