4-Fluoro-4'-methyl-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

66033-25G 66033-5G
76935-736EA 608.96 USD
76935-736 76935-738
4-Fluoro-4'-methyl-[1,1'-biphenyl]-3-carboxylic acid ≥97%
4-Fluoro-4'-methyl-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₂
MW: 230.234 g/mol
Storage Temperature: Ambient
CAS Number: 1183641-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR