5-(4-Chloro-2,6-dimethylphenyl)thiazol-2-amine ≥97%
Supplier: AOBChem USA
|
Warning
|
65283-1G
65283-500MG
65283-5G
76935-710EA
643.97
USD
76935-710
76935-712
76935-714
5-(4-Chloro-2,6-dimethylphenyl)thiazol-2-amine ≥97%
5-(4-Chloro-2,6-dimethylphenyl)thiazol-2-amine
Precaución: for Research Use Only
Formula:
C₁₁H₁₁ClN₂S Storage Temperature: Ambient |
CAS Number:
2807440-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...