5-(2,6-Difluoro-3-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

65276-1G 65276-25G 65276-500MG 65276-5G
76935-696EA 381.93 USD
76935-696 76935-698 76935-700 76935-702
5-(2,6-Difluoro-3-methoxyphenyl)thiazol-2-amine ≥97%
5-(2,6-Difluoro-3-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈F₂N₂OS
MW: 242.2451 g/mol
Storage Temperature: Ambient
CAS Number: 2807440-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR