4-Ethoxy-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65020-1G 65020-25G 65020-500MG 65020-5G
76935-682EA 322.52 USD
76935-682 76935-684 76935-686 76935-688
4-Ethoxy-3-hydroxybenzaldehyde ≥97%
4-Ethoxy-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Storage Temperature: Refrigerator
CAS Number: 2539-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR