4-Bromo-2-fluoro-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

64669-25G 64669-5G
76935-624EA 251.44 USD
76935-624 76935-626
4-Bromo-2-fluoro-6-methylaniline ≥97%
4-Bromo-2-fluoro-6-methylaniline
Formula: C₇H₇BrFN
Storage Temperature: Ambient
MDL Number: MFCD22683228
CAS Number: 429683-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR