4-Bromo-3-fluoro-2-iodophenol ≥97%

Supplier: AOBChem USA

Warning

63802-1G 63802-25G 63802-500MG 63802-5G
76935-594EA 406.33 USD
76935-594 76935-596 76935-598 76935-600
4-Bromo-3-fluoro-2-iodophenol ≥97%
4-Bromo-3-fluoro-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃BrFIO
Storage Temperature: Ambient
CAS Number: 1807008-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR