5-Bromo-4-chloro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-carboxy-5-chloroanisole, 5-Bromo-4-chloro-o-anisic acid

63585-25G 63585-5G
76935-582EA 559.1 USD
76935-582 76935-584
5-Bromo-4-chloro-2-methoxybenzoic acid ≥97%
5-Bromo-4-chloro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₃
MW: 265.488 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878363
CAS Number: 95383-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR