5-(3-Isopropoxy-4-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

63362-1G 63362-25G 63362-500MG 63362-5G
76935-538EA 334.19 USD
76935-538 76935-540 76935-542 76935-544
5-(3-Isopropoxy-4-methylphenyl)thiazol-2-amine ≥97%
5-(3-Isopropoxy-4-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂OS
MW: 248.3439 g/mol
Storage Temperature: Ambient
CAS Number: 2807445-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR