5-Bromo-4-fluoro-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

63097-1G 63097-25G 63097-500MG 63097-5G
76935-512EA 381.93 USD
76935-512 76935-514 76935-516 76935-518
5-Bromo-4-fluoro-2-methylbenzoic acid ≥95%
5-Bromo-4-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
CAS Number: 1427416-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR