5-(2-Aminothiazol-5-yl)-2-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

62699-1G 62699-25G 62699-500MG 62699-5G
76935-492EA 263.11 USD
76935-492 76935-494 76935-496 76935-498
5-(2-Aminothiazol-5-yl)-2-methoxyphenol ≥97%
5-(2-Aminothiazol-5-yl)-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂S
MW: 222.2636 g/mol
Storage Temperature: Ambient
CAS Number: 2764730-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR