5-(4-(Benzyloxy)-2-chlorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62692-1G 62692-500MG 62692-5G
76935-478EA 593.05 USD
76935-478 76935-480 76935-482
5-(4-(Benzyloxy)-2-chlorophenyl)thiazol-2-amine ≥97%
5-(4-(Benzyloxy)-2-chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₃ClN₂OS
MW: 316.8052 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR