5-(4-Bromo-3-chloro-2-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62537-1G 62537-500MG 62537-5G
76935-452EA 554.86 USD
76935-452 76935-454 76935-456
5-(4-Bromo-3-chloro-2-fluorophenyl)thiazol-2-amine ≥97%
5-(4-Bromo-3-chloro-2-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅BrClFN₂S
MW: 307.5698 g/mol
Storage Temperature: Ambient
CAS Number: 1690605-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR