5-(5-Bromo-3-chloro-2-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62535-1G 62535-500MG 62535-5G
76935-440EA 560.16 USD
76935-440 76935-442 76935-444
5-(5-Bromo-3-chloro-2-fluorophenyl)thiazol-2-amine ≥97%
5-(5-Bromo-3-chloro-2-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅BrClFN₂S
MW: 307.5698 g/mol
Storage Temperature: Ambient
CAS Number: 2764731-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR