5-([1,1'-Biphenyl]-2-yl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62532-1G 62532-25G 62532-500MG 62532-5G
76935-426EA 359.65 USD
76935-426 76935-428 76935-430 76935-432
5-([1,1'-Biphenyl]-2-yl)thiazol-2-amine ≥97%
5-([1,1'-Biphenyl]-2-yl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂S
MW: 252.3341 g/mol
Storage Temperature: Ambient
CAS Number: 2764731-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR