5-(3-Bromo-2,6-difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62530-1G 62530-500MG 62530-5G
76935-414EA 525.15 USD
76935-414 76935-416 76935-418
5-(3-Bromo-2,6-difluorophenyl)thiazol-2-amine ≥97%
5-(3-Bromo-2,6-difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅BrF₂N₂S
MW: 291.1152 g/mol
Storage Temperature: Ambient
CAS Number: 1698004-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR