5-(2-Fluoro-5-(methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62527-1G 62527-500MG 62527-5G
76935-402EA 527.27 USD
76935-402 76935-404 76935-406
5-(2-Fluoro-5-(methylthio)phenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-5-(methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S₂
MW: 240.3202 g/mol
Storage Temperature: Ambient
CAS Number: 2764730-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR