5-(2-Bromo-6-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61989-1G 61989-500MG 61989-5G
76935-380EA 494.38 USD
76935-380 76935-382 76935-384
5-(2-Bromo-6-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%
5-(2-Bromo-6-(trifluoromethoxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂OS
MW: 339.1316 g/mol
Storage Temperature: Ambient
CAS Number: 2764729-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR