5-(5-Bromo-2-fluoro-3-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61987-1G 61987-500MG 61987-5G
76935-368EA 526.21 USD
76935-368 76935-370 76935-372
5-(5-Bromo-2-fluoro-3-methylphenyl)thiazol-2-amine ≥97%
5-(5-Bromo-2-fluoro-3-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈BrFN₂S
MW: 287.1513 g/mol
Storage Temperature: Ambient
CAS Number: 2764729-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR