4-Bromo-3-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61891-1G 61891-25G 61891-5G
76935-328EA 94.43 USD
76935-328 76935-330 76935-332
4-Bromo-3-isopropylbenzoic acid ≥97%
4-Bromo-3-isopropylbenzoic acid
Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15833351
CAS Number: 741698-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR