5-(3-Chloro-5-(methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61142-1G 61142-500MG 61142-5G
76935-304EA 637.61 USD
76935-304 76935-306 76935-308
5-(3-Chloro-5-(methylthio)phenyl)thiazol-2-amine ≥97%
5-(3-Chloro-5-(methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂S₂
MW: 256.7748 g/mol
Storage Temperature: Ambient
CAS Number: 2764729-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR