5-(4-(Benzyloxy)-3-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61140-1G 61140-25G 61140-500MG 61140-5G
76935-288EA 352.22 USD
76935-288 76935-290 76935-292 76935-294
5-(4-(Benzyloxy)-3-fluorophenyl)thiazol-2-amine ≥97%
5-(4-(Benzyloxy)-3-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₃FN₂OS
MW: 300.3506 g/mol
Storage Temperature: Ambient
CAS Number: 2757730-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR