5-(4-Isopropoxy-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61138-1G 61138-500MG 61138-5G
76935-276EA 496.51 USD
76935-276 76935-278 76935-280
5-(4-Isopropoxy-2-methylphenyl)thiazol-2-amine ≥97%
5-(4-Isopropoxy-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂OS
MW: 248.3439 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR