5-(4-Butoxy-3-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61132-1G 61132-500MG 61132-5G
76935-238EA 461.5 USD
76935-238 76935-240 76935-242
5-(4-Butoxy-3-methoxyphenyl)thiazol-2-amine ≥97%
5-(4-Butoxy-3-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₂S
MW: 278.3699 g/mol
Storage Temperature: Ambient
CAS Number: 2764733-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR