5-(4-Bromo-2-isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61124-1G 61124-500MG 61124-5G
76935-224EA 702.32 USD
76935-224 76935-226 76935-228
5-(4-Bromo-2-isopropoxyphenyl)thiazol-2-amine ≥97%
5-(4-Bromo-2-isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂OS
MW: 313.2134 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR