5-(2-Fluoro-3-iodophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61121-1G 61121-500MG 61121-5G
76935-212EA 464.68 USD
76935-212 76935-214 76935-216
5-(2-Fluoro-3-iodophenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-3-iodophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆FIN₂S
Storage Temperature: Ambient
CAS Number: 2764731-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR