5-(4-Bromo-3-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61118-1G 61118-500MG 61118-5G
76935-200EA 494.38 USD
76935-200 76935-202 76935-204
5-(4-Bromo-3-methoxyphenyl)thiazol-2-amine ≥97%
5-(4-Bromo-3-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂OS
MW: 285.1602 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR