4-Bromo-N-2-dimethylaniline ≥95% research grade

Supplier: AOBChem USA

Warning

61010-1G 61010-25G 61010-5G
76935-188EA 72.15 USD
76935-188 76935-190 76935-192
4-Bromo-N-2-dimethylaniline ≥95% research grade
4-Bromo-N,2-dimethylaniline
Formula: C₈H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD11151443
CAS Number: 59557-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR