5-(3,6-Difluoro-2-methoxyphenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

60977-1G 60977-25G 60977-500MG 60977-5G
76935-172EA 381.93 USD
76935-172 76935-174 76935-176 76935-178
5-(3,6-Difluoro-2-methoxyphenyl)thiazol-2-amine ≥95%
5-(3,6-Difluoro-2-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈F₂N₂OS
MW: 242.2451 g/mol
Storage Temperature: Ambient
CAS Number: 1876680-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR