5-(3-Ethoxy-4-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

60968-1G 60968-500MG 60968-5G
76935-120EA 530.45 USD
76935-120 76935-122 76935-124
5-(3-Ethoxy-4-methylphenyl)thiazol-2-amine ≥97%
5-(3-Ethoxy-4-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂OS
MW: 234.3173 g/mol
Storage Temperature: Ambient
CAS Number: 2755717-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR