5-Bromo-2-methylsulfonylpyridine ≥97%

Supplier: AOBChem USA

Warning

60752-1G 60752-25G 60752-5G
76935-106EA 78.51 USD
76935-106 76935-108 76935-110
5-Bromo-2-methylsulfonylpyridine ≥97%
5-Bromo-2-methylsulfonylpyridine
Formula: C₆H₆BrNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD04974510
CAS Number: 98626-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR