4-Ethoxy-2-iodo-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60222-1G 60222-25G 60222-500MG 60222-5G
76935-080EA 225.98 USD
76935-080 76935-082 76935-084 76935-086
4-Ethoxy-2-iodo-1-methylbenzene ≥97%
4-Ethoxy-2-iodo-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO
MW: 262.088 g/mol
Storage Temperature: Ambient
CAS Number: 2764731-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR