4-Fluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

59989-25G 59989-5G
76935-054EA 233.4 USD
76935-054 76935-056
4-Fluoro-3-formylbenzoic acid ≥97%
4-Fluoro-3-formylbenzoic acid
Formula: C₈H₅FO₃
MW: 168.1200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06411555
CAS Number: 845885-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR