4-Bromo-2-isopropoxy-1-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

59906-1G 59906-25G 59906-500MG 59906-5G
76935-040EA 203.7 USD
76935-040 76935-042 76935-044 76935-046
4-Bromo-2-isopropoxy-1-(trifluoromethoxy)benzene ≥95%
4-Bromo-2-isopropoxy-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrF₃O₂
MW: 299.084 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR