4-Butoxy-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

59726-1G 59726-25G 59726-5G
76935-026EA 81.69 USD
76935-026 76935-028 76935-030
4-Butoxy-2-fluorobenzoic acid ≥97%
4-Butoxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
CAS Number: 123843-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR