5-(3-Ethoxy-4-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59550-1G 59550-500MG 59550-5G
76935-000EA 563.34 USD
76935-000 76935-002 76935-004
5-(3-Ethoxy-4-fluorophenyl)thiazol-2-amine ≥97%
5-(3-Ethoxy-4-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁FN₂OS
MW: 238.281 g/mol
Storage Temperature: Ambient
CAS Number: 2755718-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR