5-(3-Chloro-2-ethoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59545-1G 59545-500MG 59545-5G
76934-988EA 663.07 USD
76934-988 76934-990 76934-992
5-(3-Chloro-2-ethoxyphenyl)thiazol-2-amine ≥97%
5-(3-Chloro-2-ethoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂OS
Storage Temperature: Ambient
CAS Number: 2755722-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR