5-(4-Isopropoxy-3-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59542-1G 59542-500MG
76934-976EA 495.45 USD
76934-976 76934-978
5-(4-Isopropoxy-3-methoxyphenyl)thiazol-2-amine ≥97%
5-(4-Isopropoxy-3-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂S
MW: 264.343 g/mol
Storage Temperature: Ambient
CAS Number: 2755717-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR