5-(3-Methoxy-5-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59540-1G 59540-500MG 59540-5G
76934-962EA 500.75 USD
76934-962 76934-964 76934-966
5-(3-Methoxy-5-methylphenyl)thiazol-2-amine ≥97%
5-(3-Methoxy-5-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
MW: 220.291 g/mol
Storage Temperature: Ambient
CAS Number: 2755718-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR