5-(3-Chloro-4-ethoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59538-1G 59538-25G 59538-500MG 59538-5G
76934-948EA 441.34 USD
76934-948 76934-950 76934-952 76934-954
5-(3-Chloro-4-ethoxyphenyl)thiazol-2-amine ≥97%
5-(3-Chloro-4-ethoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂OS
Storage Temperature: Ambient
CAS Number: 2755720-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR