5-(3-(Benzyloxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59536-1G 59536-25G 59536-500MG 59536-5G
76934-934EA 375.56 USD
76934-934 76934-936 76934-938 76934-940
5-(3-(Benzyloxy)phenyl)thiazol-2-amine ≥97%
5-(3-(Benzyloxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂OS
MW: 282.36 g/mol
Storage Temperature: Ambient
CAS Number: 1995660-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR