5-(5-Chloro-2-(methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59534-1G 59534-500MG 59534-5G
76934-922EA 671.55 USD
76934-922 76934-924 76934-926
5-(5-Chloro-2-(methylthio)phenyl)thiazol-2-amine ≥97%
5-(5-Chloro-2-(methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂S₂
MW: 256.775 g/mol
Storage Temperature: Ambient
CAS Number: 2755722-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR