5-(4-Chloro-3-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59530-1G 59530-25G 59530-500MG 59530-5G
76934-896EA 441.34 USD
76934-896 76934-898 76934-900 76934-902
5-(4-Chloro-3-methoxyphenyl)thiazol-2-amine ≥97%
5-(4-Chloro-3-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂OS
MW: 240.709 g/mol
Storage Temperature: Ambient
CAS Number: 1538443-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR