5-(2-Ethoxy-5-fluorophenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

59417-1G 59417-25G 59417-500MG 59417-5G
76934-868EA 396.78 USD
76934-868 76934-870 76934-872 76934-874
5-(2-Ethoxy-5-fluorophenyl)thiazol-2-amine ≥95%
5-(2-Ethoxy-5-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁FN₂OS
MW: 238.281 g/mol
Storage Temperature: Ambient
CAS Number: 2755717-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR