5-(2-Fluoro-3-propoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59413-1G 59413-25G 59413-500MG 59413-5G
76934-852EA 434.97 USD
76934-852 76934-854 76934-856 76934-858
5-(2-Fluoro-3-propoxyphenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-3-propoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃FN₂OS
MW: 252.308 g/mol
Storage Temperature: Ambient
CAS Number: 2755723-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR