5-(3-Isopropyl-5-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59408-1G 59408-500MG 59408-5G
76934-838EA 725.66 USD
76934-838 76934-840 76934-842
5-(3-Isopropyl-5-methylphenyl)thiazol-2-amine ≥97%
5-(3-Isopropyl-5-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂S
MW: 232.345 g/mol
Storage Temperature: Ambient
CAS Number: 2755724-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR