5-(4-Pentylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59406-1G 59406-25G 59406-500MG 59406-5G
76934-824EA 396.78 USD
76934-824 76934-826 76934-828 76934-830
5-(4-Pentylphenyl)thiazol-2-amine ≥97%
5-(4-Pentylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂S
MW: 246.371 g/mol
Storage Temperature: Ambient
CAS Number: 2755723-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR